2,5-Dibromo-3-dodecylthiophene 98%

Supplier: Ambeed
Warning

A166183-1G A166183-5G
103806-018EA 10 USD
103806-018 103844-278 77320-514 77320-516
2,5-Dibromo-3-dodecylthiophene 98%
2,5-Dibromo-3-dodecylthiophene

Precaución: for Research Use Only

Formula: C₁₆H₂₆Br₂S
Boiling Pt: 407.0±40.0 °C (760 mm Hg)
Flash Pt: 199.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00274315
CAS Number: 148256-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR