2,5-Dibromo-3-fluorochlorobenzene 97%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dibromo-1-chloro-3-fluorobenzene

A256486-10G A256486-1G A256486-5G
103812-270EA 53 USD
103812-270 103852-134 76874-718 77091-630 77304-678
2,5-Dibromo-3-fluorochlorobenzene 97%
2,5-Dibromo-3-fluorochlorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂ClF
MW: 288.35 g/mol
Boiling Pt: 248.3 °C (760 mm Hg)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD09878162
CAS Number: 1000572-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR