2,5-Dibromo-m-xylene 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dibromo-1,3-dimethylbenzene, 1,4-Dibromo-2,6-dimethylbenzene

A183233-100G A183233-10G A183233-1G
103815-622EA 6 USD
103815-622 103846-184 77248-332 77248-334 77301-830
2,5-Dibromo-m-xylene 98%
2,5-Dibromo-m-xylene

Precaución: for Research Use Only

Formula: C₈H₈Br₂
MW: 263.96
Storage Temperature: Ambient
MDL Number: MFCD00079745
CAS Number: 100189-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR