2,5-Diaminopyrimidine-4,6-diol 95%

Supplier: Ambeed
Warning

A229770-1G A229770-250MG
103819-492EA 29 USD
103819-492 103850-394 77089-242 77193-096
2,5-Diaminopyrimidine-4,6-diol 95%
2,5-Diaminopyrimidine-4,6-diol

Precaución: for Research Use Only

Formula: C₄H₆N₄O₂
MW: 142.12 g/mol
Boiling Pt: 524.2 °C (760 mm Hg)
Melting Pt: >250 °C
Flash Pt: 270.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02094729
CAS Number: 40769-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR