2,5-Diamino-4,6-dichloropyrimidine 98%

Supplier: Ambeed
Warning

A134785-100G A134785-10G A134785-1G A134785-25G A134785-5G
103802-050EA 13 USD
103802-050 77285-464 77285-466 77314-822 77314-824 77314-826
2,5-Diamino-4,6-dichloropyrimidine 98%
2,5-Diamino-4,6-dichloropyrimidine

Precaución: for Research Use Only

Formula: C₄H₄Cl₂N₄
Boiling Pt: 395.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00792652
CAS Number: 55583-59-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR