α,α,α,α',α',α'-Hexafluoro-2,5-xylidine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,5-Bis(trifluoromethyl)aniline
A170198-100G
A170198-10G
A170198-1G
A170198-500G
A170198-25G
A170198-5G
103844-726EA
6
USD
103844-726
77236-242
77236-244
77236-246
77236-248
77301-028
77301-030
α,α,α,α',α',α'-Hexafluoro-2,5-xylidine 98%
α,α,α,α',α',α'-Hexafluoro-2,5-xylidine
Precaución: for Research Use Only
Formula:
C₈H₅F₆N MW: 229.12 g/mol Boiling Pt: 177.9 °C (760 mm Hg) Flash Pt: 71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00074940 CAS Number: 328-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...