Thiophene-2,5-diboronic acid bis(pinacol) ester 97%

Supplier: Ambeed
Warning

Synonyms: 2,5-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene, Thiophene-2,5-diboronic acid Bis(pinacol) ester, 2,2'-(2,5-Thiophenediyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)

A292442-1G A292442-25G A292442-100G A292442-10G A292442-250MG
103813-940EA 7 USD
103813-940 103835-080 76837-682 76837-684 77014-816 77014-818 77014-820
Thiophene-2,5-diboronic acid bis(pinacol) ester 97%
Thiophene-2,5-diboronic acid bis(pinacol) ester

Precaución: for Research Use Only

Formula: C₁₆H₂₆B₂O₄S
MW: 336.06 g/mol
Boiling Pt: 434.3 °C (760 mm Hg)
Melting Pt: 227…231 °C
Flash Pt: 216.5 °C
Storage Temperature: Freezer
MDL Number: MFCD09800563
CAS Number: 175361-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR