2,5-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thieno[3,2-b]thiophene 97%
Supplier: Ambeed
|
Warning
|
A200584-100MG
A200584-250MG
A200584-1G
A200584-10G
103817-214EA
263
USD
103817-214
103847-846
76873-382
76873-384
77302-692
77611-944
2,5-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thieno[3,2-b]thiophene 97%
2,5-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thieno[3,2-b]thiophene
Precaución: for Research Use Only
Formula:
C₁₈H₂₆B₂O₄S₂ MW: 392.14 g/mol Boiling Pt: 501.0±45.0 °C (760 mm Hg) Melting Pt: 290…294 °C Flash Pt: 256.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD22200472 CAS Number: 924894-85-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...