Hexaethylene glycol monomethyl ether 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
mPEG6-Alcohol, 2,5,8,11,14,17-Hexaoxanonadecan-19-ol
A117126-250MG
A117126-1G
A117126-5G
A117126-25G
A117126-100G
103798-832EA
13
USD
103798-832
103824-708
76797-120
77311-490
77311-492
77549-346
77666-684
Hexaethylene glycol monomethyl ether 98%
Hexaethylene glycol monomethyl ether
Precaución: for Research Use Only
Formula:
C₁₃H₂₈O₇ MW: 296.36 g/mol Boiling Pt: 375.6 °C (760 mm Hg) Flash Pt: 180.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00045996 CAS Number: 23601-40-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...