2,5,6-Trichloronicotinonitrile 95%

Supplier: Ambeed
Warning

A257508-1G A257508-250MG
103812-312EA 21 USD
103812-312 103852-178 77091-686 77672-296
2,5,6-Trichloronicotinonitrile 95%
2,5,6-Trichloronicotinonitrile

Precaución: for Research Use Only

Formula: C₆HCl₃N₂
MW: 207.45 g/mol
Boiling Pt: 285.6±35.0 °C (760 mm Hg)
Melting Pt: 118…121 °C
Flash Pt: 126.5 °C
Storage Temperature: Ambient
MDL Number: MFCD17392884
CAS Number: 40381-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR