2,5,6-Trichloronicotinic acid 97%

Supplier: Ambeed
Warning

A501134-100G A501134-10G A501134-250MG A501134-25G A501134-1G A501134-5G
103828-292EA 12 USD
103828-292 103839-380 76830-198 76830-200 76830-202 76830-204 76843-340 76843-342
2,5,6-Trichloronicotinic acid 97%
2,5,6-Trichloronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₂Cl₃NO₂
MW: 226.45 g/mol
Boiling Pt: 359.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD02070018
CAS Number: 54718-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR