2,4-Hexanedione 97%
Supplier: Ambeed
|
Danger
|
A522757-1G
A522757-5G
A522757-250MG
103828-518EA
10
USD
103828-518
103839-632
76843-632
76843-634
77253-518
2,4-Hexanedione 97%
2,4-Hexanedione
Precaución: for Research Use Only
Formula:
C₆H₁₀O₂ Boiling Pt: 161 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009318 CAS Number: 3002-24-2 UN: 1224 ADR: 3,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...