2,4-Di-tert-butylaniline 98%

Supplier: Ambeed
Warning

A168104-100MG A168104-250MG A168104-1G
103806-250EA 288 USD
103806-250 103844-508 77096-422 77096-424 77300-918
2,4-Di-tert-butylaniline 98%
2,4-Di-tert-butylaniline

Precaución: for Research Use Only

Formula: C₁₄H₂₃N
Storage Temperature: Ambient
MDL Number: MFCD02683807
CAS Number: 2909-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR