2,4-Dimethylpyrimidin-5-ol 98%

Supplier: Ambeed
Warning

A153597-250MG A153597-100MG A153597-10G
103804-534EA 94 USD
103804-534 103842-708 77611-452 77671-150 77671-152
2,4-Dimethylpyrimidin-5-ol 98%
2,4-Dimethylpyrimidin-5-ol

Precaución: Inert atmosphere, room temperature

Formula: C₆H₈N₂O
Boiling Pt: 219.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD18072447
CAS Number: 412003-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR