2,4-Dimethylphenylboronic acid 98%

Supplier: Ambeed
Warning

A165802-100G A165802-10G A165802-1G A165802-25G A165802-5G
103806-000EA 6 USD
103806-000 103844-254 77191-250 77191-252 77320-474 77320-476 77320-478
2,4-Dimethylphenylboronic acid 98%
2,4-Dimethylphenylboronic acid

Precaución: Keep in a dark place, sealed in dry, room temperature

Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
Boiling Pt: 292.4 °C (760 mm Hg)
Melting Pt: 150 °C
Flash Pt: 130.7 °C
Storage Temperature: Ambient
MDL Number: MFCD02683101
CAS Number: 55499-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR