6-Nitro-2,4-xylidine 97%

Supplier: Ambeed
Warning

Synonyms: 4,6-Dimethyl-2-nitroaniline, 1-Amino-2,4-dimethyl-6-nitrobenzene, (2,4-Dimethyl-6-nitrophenyl)amine, 2,4-Dimethyl-6-nitroaniline

A100473-10G A100473-1G
103795-460EA 7 USD
103795-460 103821-154 77188-522 77296-702
6-Nitro-2,4-xylidine 97%
6-Nitro-2,4-xylidine

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Boiling Pt: 319.1 °C (760 mm Hg)
Melting Pt: 64…68 °C
Storage Temperature: Freezer
MDL Number: MFCD00007739
CAS Number: 1635-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR