5-Nitro-2,4-xylidine 97%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dimethyl-5-nitroaniline, 5-Nitro-2,4-xylidine

A105979-10G A105979-250MG A105979-25G A105979-1G A105979-5G
103796-978EA 14 USD
103796-978 103822-728 77245-432 77245-434 77245-436 77309-588 77309-590
5-Nitro-2,4-xylidine 97%
5-Nitro-2,4-xylidine

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.1800 g/mol
Boiling Pt: 329.4 °C (760 mm Hg)
Flash Pt: 153 °C
Storage Temperature: Ambient
MDL Number: MFCD00025199
CAS Number: 2124-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR