2,4-Dimethyl-1H-imidazole-5-carbaldehyde 95%

Supplier: Ambeed
Warning

A278253-100MG A278253-250MG
103813-406EA 133 USD
103813-406 103853-348 76827-468 76827-470
2,4-Dimethyl-1H-imidazole-5-carbaldehyde 95%
2,4-Dimethyl-1H-imidazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MW: 124.14 g/mol
Boiling Pt: 365.377 °C (760 mm Hg)
Melting Pt: 159…161 °C
Flash Pt: 178.437 °C
Storage Temperature: Refrigerator
MDL Number: MFCD19300742
CAS Number: 68282-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR