2,4-Dimethoxy-5-pyrimidineboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dimethoxypyrimidine-5-boronic acid, (2,4-Dimethoxypyrimidin-5-yl)boronic acid, 2,4-Dimethoxy-5-pyrimidylboronic acid

A179168-1G A179168-25G A179168-5G A179168-100G A179168-10G A179168-250MG
103807-406EA 16 USD
103807-406 103845-752 77083-206 77083-208 77083-210 77111-346 77111-348 77111-350
2,4-Dimethoxy-5-pyrimidineboronic acid 97%
2,4-Dimethoxy-5-pyrimidineboronic acid

Precaución: for Research Use Only

Formula: C₆H₉BN₂O₄
MW: 183.9577 g/mol
Boiling Pt: 420.6 °C (760 mm Hg)
Melting Pt: 113…117 °C
Flash Pt: 208.1 °C
Storage Temperature: Freezer
MDL Number: MFCD01318985
CAS Number: 89641-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR