2,4-Dimethoxypyrimidine 98%

Supplier: Ambeed
Warning

A206714-25G A206714-5G A206714-100G A206714-1G
103817-726EA 5 USD
103817-726 103848-402 77086-660 77086-662 77112-340 77303-000
2,4-Dimethoxypyrimidine 98%
2,4-Dimethoxypyrimidine

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
Boiling Pt: 241.6 °C (760 mm Hg)
Melting Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00056028
CAS Number: 3551-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR