2,4-Dimethoxypyrimidin-5-amine 95%

Supplier: Ambeed
Warning

A203765-100MG A203765-250MG A203765-1G A203765-5G
103817-488EA 382 USD
103817-488 103848-148 77012-732 77012-734 77302-868 77302-870
2,4-Dimethoxypyrimidin-5-amine 95%
2,4-Dimethoxypyrimidin-5-amine

Precaución: for Research Use Only

Formula: C₆H₉N₃O₂
Melting Pt: 89 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12546337
CAS Number: 14048-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR