2,4-Dimethoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

A446974-25G A446974-5G A446974-100G A446974-1G A446974-500G
103827-750EA 9 USD
103827-750 103838-816 76842-620 76842-622 77196-436 77307-346 77616-926
2,4-Dimethoxyphenylboronic acid 98%
2,4-Dimethoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₄
MW: 181.98 g/mol
Boiling Pt: 361.1 °C (760 mm Hg)
Melting Pt: 125…127 °C
Flash Pt: 172.2 °C
Storage Temperature: Ambient
MDL Number: MFCD01074590
CAS Number: 133730-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR