2,4-Dihydroxy-5-isopropylbenzoic acid 95%

Supplier: Ambeed
Warning

A130543-100MG A130543-250MG A130543-1G A130543-5G
103801-434EA 88 USD
103801-434 77149-574 77149-576 77298-412 77549-546
2,4-Dihydroxy-5-isopropylbenzoic acid 95%
2,4-Dihydroxy-5-isopropylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
MW: 196.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16996083
CAS Number: 1184181-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR