2,4-Dihydroxy-5,6-dimethylpyrimidine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5,6-Dimethyl-2,4-pyrimidinediol, 5,6-Dimethyluracil, 2,4-Dihydroxy-5,6-dimethylpyrimidine
A127618-10G
A127618-250MG
A127618-1G
103800-988EA
12
USD
103800-988
76766-958
76766-960
77313-234
2,4-Dihydroxy-5,6-dimethylpyrimidine 97%
2,4-Dihydroxy-5,6-dimethylpyrimidine
Precaución: for Research Use Only
Formula:
C₆H₈N₂O₂ MW: 140.1400 g/mol Boiling Pt: 408.8 °C (760 mm Hg) Melting Pt: 297…300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010708 CAS Number: 26305-13-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...