4-Hydroxy-3-nitropyridin-2(1H)-one 98%

Supplier: Ambeed
Warning

A264497-100G A264497-10G A264497-5G
103852-574EA 9 USD
103852-574 77250-210 77250-212 77304-896
4-Hydroxy-3-nitropyridin-2(1H)-one 98%
4-Hydroxy-3-nitropyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₅H₄N₂O₄
MW: 156.10 g/mol
Melting Pt: 265 °C
Flash Pt: 110.6 °C
Storage Temperature: Ambient
MDL Number: MFCD20278218
CAS Number: 89282-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR