2,4-Dihydroxy-3-methylbenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A124534-100G
A124534-10G
A124534-250MG
A124534-25G
A124534-1G
A124534-5G
103800-540EA
7
USD
103800-540
76810-738
76810-740
76810-742
76810-744
77312-676
77312-678
2,4-Dihydroxy-3-methylbenzaldehyde 98%
2,4-Dihydroxy-3-methylbenzaldehyde
Precaución: for Research Use Only
Formula:
C₈H₈O₃ Boiling Pt: 345.7 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD00156883 CAS Number: 6248-20-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...