2,4-Difluorophenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
A125233-100G
A125233-10G
A125233-25G
A125233-500G
A125233-5G
A125233-1G
103800-646EA
6
USD
103800-646
77285-082
77285-084
77285-086
77285-088
77298-038
77312-794
2,4-Difluorophenylboronic acid 97%
2,4-Difluorophenylboronic acid
Precaución: for Research Use Only
Formula:
C₆H₅BF₂O₂ Melting Pt: 247…250 °C Flash Pt: 105.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01318998 CAS Number: 144025-03-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...