2,4-Difluorobenzaldehyde 96%

Supplier: Ambeed
Danger

A285186-10G A285186-25G A285186-5G
103834-794EA 5 USD
103834-794 77099-236 77305-468 77305-470
2,4-Difluorobenzaldehyde 96%
2,4-Difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 168.6 °C (760 mm Hg)
Melting Pt: 2…3 °C
Flash Pt: 55 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010326
CAS Number: 1550-35-2
UN: 1989
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR