2,4-Difluoro-5-nitroaniline 98%
Supplier: Ambeed
|
Warning
|
A285654-1G
A285654-5G
A285654-100G
A285654-100MG
A285654-10G
A285654-250MG
A285654-25G
103813-684EA
16
USD
103813-684
103834-818
76837-354
76837-356
76988-562
76988-564
76988-566
76988-568
76988-570
2,4-Difluoro-5-nitroaniline 98%
2,4-Difluoro-5-nitroaniline
Precaución: for Research Use Only
Formula:
C₆H₄F₂N₂O₂ MW: 174.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04038395 CAS Number: 123344-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...