2,4-Difluorotoluene 97%

Supplier: Ambeed
Danger

Synonyms: 2,4-Difluoro-1-methylbenzene

A429579-5G A429579-25G
103838-564EA 6 USD
103838-564 76842-254 77572-334
2,4-Difluorotoluene 97%
2,4-Difluorotoluene

Precaución: for Research Use Only

Formula: C₇H₆F₂
MW: 128.12 g/mol
Boiling Pt: 114.7 °C (760 mm Hg)
Flash Pt: 15 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000332
CAS Number: 452-76-6
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR