2,4-Diethylthioxanthen-9-one 97%

Supplier: Ambeed
Warning

Synonyms: 2,4-Diethyl-9H-thioxanthen-9-one

A234262-100G A234262-10G A234262-500G A234262-1000G A234262-25G
103809-652EA 206 USD
103809-652 77031-140 77031-142 77031-144 77089-676 77303-974
2,4-Diethylthioxanthen-9-one 97%
2,4-Diethylthioxanthen-9-one

Precaución: for Research Use Only

Formula: C₁₇H₁₆OS
MW: 268.37 g/mol
Boiling Pt: 427.9 °C (760 mm Hg)
Melting Pt: 66…70 °C
Flash Pt: 237.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00082910
CAS Number: 82799-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR