2,4-Dichlorothieno[3,2-d]pyrimidine 95%

Supplier: Ambeed
Warning

A188460-100G A188460-10G A188460-250MG A188460-1G A188460-5G A188460-25G
103816-102EA 9 USD
103816-102 103846-698 76986-002 76986-004 76986-006 77084-440 77084-442 77302-128
2,4-Dichlorothieno[3,2-d]pyrimidine 95%
2,4-Dichlorothieno[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₂Cl₂N₂S
MW: 205.0645 g/mol
Melting Pt: 140…144 °C
Storage Temperature: Ambient
MDL Number: MFCD08448158
CAS Number: 16234-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR