2,4-Dichlorothieno[2,3-d]pyrimidine 98%

Supplier: Ambeed
Warning

A319798-1G A319798-25G A319798-5G A319798-100G A319798-10G A319798-250MG
103825-220EA 16 USD
103825-220 103836-152 76839-006 76839-008 76839-010 77002-832 77002-834 77002-836
2,4-Dichlorothieno[2,3-d]pyrimidine 98%
2,4-Dichlorothieno[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₂Cl₂N₂S
MW: 205.0645 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09743991
CAS Number: 18740-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR