2,4-Dichlorothiazole-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A227546-10G A227546-250MG A227546-1G A227546-5G
103819-324EA 176 USD
103819-324 103850-230 76771-040 76771-042 77303-800 77303-802
2,4-Dichlorothiazole-5-carboxylic acid 95%
2,4-Dichlorothiazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₄HCl₂NO₂S
MW: 198.0300 g/mol
Boiling Pt: 385 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09909676
CAS Number: 62019-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR