2,4-Dichloroquinoline 98%

Supplier: Ambeed
Warning

A277996-100G A277996-10G A277996-25G A277996-5G A277996-1G
103813-386EA 8 USD
103813-386 103853-324 76692-974 76692-976 77093-230 77093-232 77305-284
2,4-Dichloroquinoline 98%
2,4-Dichloroquinoline

Precaución: for Research Use Only

Formula: C₉H₅Cl₂N
Boiling Pt: 281 °C (760 mm Hg)
Melting Pt: 52…54 °C
Flash Pt: 154.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00023939
CAS Number: 703-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR