2,4-Dichloropyrimidine 98%
Supplier: Ambeed
|
Warning
|
A108574-100G
A108574-10G
A108574-25G
A108574-500G
A108574-5G
103807-998EA
259
USD
103807-998
77283-750
77283-752
77283-754
77283-756
77283-758
77309-990
2,4-Dichloropyrimidine 98%
2,4-Dichloropyrimidine
Precaución: for Research Use Only
Formula:
C₄H₂Cl₂N₂ Boiling Pt: 209.1 °C (760 mm Hg) Melting Pt: 57…61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006061 CAS Number: 3934-20-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...