2,4-Dichloropyrido[3,4-d]pyrimidine 96%

Supplier: Ambeed
Warning

A128105-100MG A128105-250MG
103801-070EA 242 USD
103801-070 76782-146 77666-824
2,4-Dichloropyrido[3,4-d]pyrimidine 96%
2,4-Dichloropyrido[3,4-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₃Cl₂N₃
MW: 200.025 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10699461
CAS Number: 908240-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR