2,4-Dichloropyrido[2,3-d]pyrimidine 98%

Supplier: Ambeed
Warning

A123866-10G A123866-5G A123866-1G
103800-456EA 40 USD
103800-456 77149-316 77297-962 77312-578
2,4-Dichloropyrido[2,3-d]pyrimidine 98%
2,4-Dichloropyrido[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₃Cl₂N₃
Storage Temperature: Refrigerator
MDL Number: MFCD07644627
CAS Number: 126728-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR