2,4-Dichlorophenylboronic acid 98%

Supplier: Ambeed
Warning

A163466-100G A163466-10G A163466-1G A163466-25G A163466-5G A163466-500G
103805-732EA 9 USD
103805-732 103843-992 76812-870 76812-872 77300-662 77320-090 77320-092 77671-304
2,4-Dichlorophenylboronic acid 98%
2,4-Dichlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BCl₂O₂
Boiling Pt: 331.3 °C (760 mm Hg)
Melting Pt: 246…249 °C
Flash Pt: 154.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00013930
CAS Number: 68716-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR