2,4-Dichloro-3-cyanopyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dichloropyridine-3-carbonitrile, 2,4-Dichloronicotinonitrile

A141567-10G A141567-250MG A141567-1G A141567-25G A141567-5G
103803-016EA 14 USD
103803-016 77246-974 77246-976 77316-182 77316-184 77316-186
2,4-Dichloro-3-cyanopyridine 98%
2,4-Dichloro-3-cyanopyridine

Precaución: for Research Use Only

Formula: C₆H₂Cl₂N₂
MW: 172.996 g/mol
Boiling Pt: 293 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09750302
CAS Number: 180995-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR