2,4-Dichloro-3-formylpyridine 98%
Supplier: Ambeed
|
Warning
|
A127652-100MG
A127652-10G
A127652-250MG
A127652-1G
A127652-25G
A127652-5G
103800-994EA
9
USD
103800-994
77246-376
77246-378
77246-380
77313-242
77313-244
77313-246
2,4-Dichloro-3-formylpyridine 98%
2,4-Dichloro-3-formylpyridine
Precaución: for Research Use Only
Formula:
C₆H₃Cl₂NO Boiling Pt: 261.7 °C (760 mm Hg) Melting Pt: 72…75 °C Flash Pt: 112.104 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07437909 CAS Number: 134031-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...