2,4-Dichlorofuro[3,2-d]pyrimidine 97%

Supplier: Ambeed
Warning

A311543-100MG A311543-250MG A311543-5G
103814-672EA 59 USD
103814-672 103835-864 76787-934 76787-936 76838-666
2,4-Dichlorofuro[3,2-d]pyrimidine 97%
2,4-Dichlorofuro[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₂Cl₂N₂O
MW: 188.9989 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11520866
CAS Number: 956034-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR