2,4-Dichlorocinnamic acid 97%

Supplier: Ambeed
Warning

A150945-10G A150945-5G
103842-364EA 11 USD
103842-364 77259-248 77317-898
2,4-Dichlorocinnamic acid 97%
2,4-Dichlorocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₆Cl₂O₂
MW: 217.04 g/mol
Melting Pt: 233…235 °C
Storage Temperature: Ambient
MDL Number: MFCD00004373
CAS Number: 1201-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR