2,4-Dichloro-8-methylquinazoline 97%

Supplier: Ambeed
Warning

A262923-100MG A262923-250MG A262923-1G
103812-588EA 57 USD
103812-588 103852-484 76988-210 76988-212 77092-116
2,4-Dichloro-8-methylquinazoline 97%
2,4-Dichloro-8-methylquinazoline

Precaución: for Research Use Only

Formula: C₉H₆Cl₂N₂
Boiling Pt: 292.4 °C (760 mm Hg)
Flash Pt: 158.4 °C
Storage Temperature: Ambient
MDL Number: MFCD11849266
CAS Number: 39576-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR