2,4-Dichloro-7-methylthieno[3,2-d]pyrimidine 97%

Supplier: Ambeed
Warning

A106742-100MG A106742-250MG A106742-5G A106742-1G A106742-10G
103797-068EA 27 USD
103797-068 103822-826 77025-486 77025-488 77283-508 77309-678 77599-550
2,4-Dichloro-7-methylthieno[3,2-d]pyrimidine 97%
2,4-Dichloro-7-methylthieno[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₄Cl₂N₂S
MW: 219.0900 g/mol
Boiling Pt: 293 °C (760 mm Hg)
Flash Pt: 131 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08061253
CAS Number: 35265-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR