2,4-Dichloro-7-methyl-7H-pyrrolo[2,3-d]pyrimidine 97%

Supplier: Ambeed
Warning

A116651-100G A116651-100MG A116651-250MG A116651-25G A116651-1G A116651-10G
103798-778EA 16 USD
103798-778 103824-642 76810-224 76810-226 76810-228 76810-230 77311-438 77610-954
2,4-Dichloro-7-methyl-7H-pyrrolo[2,3-d]pyrimidine 97%
2,4-Dichloro-7-methyl-7H-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₅Cl₂N₃
Storage Temperature: Refrigerator
MDL Number: MFCD13189421
CAS Number: 90213-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR