2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,6-Dichloro-7-deazapurine
A181078-25G
A181078-5G
A181078-100G
A181078-10G
A181078-1G
A181078-250MG
A181078-500G
103845-948EA
15
USD
103845-948
77083-464
77083-466
77151-604
77151-606
77151-608
77151-610
77151-612
2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine 97%
2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine
Precaución: for Research Use Only
Formula:
C₆H₃Cl₂N₃ MW: 188.01 g/mol Melting Pt: 247…250 °C Flash Pt: 142.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD08059278 CAS Number: 90213-66-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...