2,4-Dichloro-7-fluoroquinazoline 95%
Supplier: Ambeed
|
Warning
|
A148596-100MG
A148596-250MG
103803-948EA
33
USD
103803-948
103842-064
77247-174
77247-176
2,4-Dichloro-7-fluoroquinazoline 95%
2,4-Dichloro-7-fluoroquinazoline
Precaución: for Research Use Only
Formula:
C₈H₃Cl₂FN₂ MW: 217.0272 g/mol Boiling Pt: 201.7 °C (760 mm Hg) Storage Temperature: Freezer |
MDL Number:
MFCD09954885 CAS Number: 174566-15-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...