2,4-Dichloro-6-phenylpyrimidine 97%

Supplier: Ambeed
Warning

A174177-100MG A174177-250MG A174177-1G A174177-5G A174177-25G
103806-930EA 28 USD
103806-930 103845-228 76985-568 76985-570 77082-522 77082-524 77301-324
2,4-Dichloro-6-phenylpyrimidine 97%
Formula: C₁₀H₆Cl₂N₂
Boiling Pt: 361.2 °C (760 mm Hg)
Flash Pt: 203.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00234591
CAS Number: 26032-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR