2,4-Dichloro-6-methylthieno[3,2-d]pyrimidine 95%

Supplier: Ambeed
Warning

A158258-100MG A158258-10G A158258-250MG A158258-1G A158258-5G
103805-112EA 29 USD
103805-112 103843-320 76812-574 76812-576 76812-578 77319-174 77319-176
2,4-Dichloro-6-methylthieno[3,2-d]pyrimidine 95%
2,4-Dichloro-6-methylthieno[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₄Cl₂N₂S
Boiling Pt: 293.1±22.0 °C (760 mm Hg)
Flash Pt: 131.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10000842
CAS Number: 35265-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR